Learn More
Electron Microscopy Sciences Microtome Thick Embedding Stubs, 10 mm
SDP
Catalog No. 5034085
Encompass_Preferred
Block holder molded from hard plastic. Easy to store block holders attached to Aluminum chucks (ABH). Frosted surface for easy marking. Large center hole prevents bubble formation.
Size: 10 mm
Catalog No. 50-340-85 Supplier Electron Microscopy Sciences Supplier No. 7017560
Block holder molded from hard plastic. Easy to store block holders attached to Aluminum chucks (ABH). Frosted surface for easy marking. Large center hole prevents bubble formation.
Size: 10 mm