Learn More
Electron Microscopy Sciences EBH-2 Microtome Block Holder, 6 mm
SDP
Catalog No. 5034089
Encompass_Preferred
Block holder molded from hard plastic. Easy to store block holders attached to Aluminum chucks (ABH). Frosted surface for easy marking. Large center hole prevents bubble formation.
Size: 6 mm
Catalog No. 50-340-89 Supplier Electron Microscopy Sciences Supplier No. 7017550
Block holder molded from hard plastic. Easy to store block holders attached to Aluminum chucks (ABH). Frosted surface for easy marking. Large center hole prevents bubble formation.
Size: 6 mm