Learn More
Fine Science Tools STUDENT DUMONT FORCEPS CURVED
SDP
Catalog No. NC9420074
Encompass
Style: #7/ Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Inox/ Length: 11cm/ Feature: Student Quality
Catalog No. NC9420074 Supplier Fine Science Tools Supplier No. 9119700
Style: #7/ Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Inox/ Length: 11cm/ Feature: Student Quality