Learn More
GSC Go Science Crazy Electrolysis Kit with Telescoping Tubes and Support
SCI_ED
Catalog No. S43482 Shop All GSC Products
Click to view available options
Product Type:
Electrolysis Kit
Catalog No. Product Type
S43482 Electrolysis Kit
1 options
Not a toy. Product intended for ages 13 and up. May contain latex.
Catalog No. S43482 Supplier GSC Go Science Crazy Supplier No. 501
Not a toy. Product intended for ages 13 and up. May contain latex.
Designed for student experiments in electrolysis.
  • Consists of a H-shaped glass tube fitted with graduated gas collection tubes
  • Includes 2 platinum electrodes
  • Cast iron support stand and clamp
  • H-tube is 22 O.D. x 150mm L x 75mm W
  • Collecting tubes is 14mm O.D. x 150mm L, graduated to 14mm
  • Collecting tubes fitted with pair of high quality platinum electrodes
Specifications
Includes Two telescoping gas-collecting tube and platinum electrodes
Product Type Electrolysis Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov