Learn More
Fine Science Tools Student Fine Forceps, Stainless Steel, Angled, 11 cm
SDP
Catalog No. NC0991373
Encompass
Tip Shape: Angled/ Tips: Standard/ Tip Dimensions: 0.2 x 0.2mm/ Alloy / Material: Student Stainless Steel/ Length: 11cm/ Feature: Student Quality
Catalog No. NC0991373 Supplier Fine Science Tools Supplier No. 9111010
Tip Shape: Angled/ Tips: Standard/ Tip Dimensions: 0.2 x 0.2mm/ Alloy / Material: Student Stainless Steel/ Length: 11cm/ Feature: Student Quality