Learn More
Fine Science Tools Student Fine Forceps, Curved, Student Stainless Steel, 12 cm
SDP
Catalog No. NC1130608
Encompass
Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.2 x 0.2mm/ Alloy / Material: Student Stainless Steel/ Length: 11cm/ Feature: Student Quality
Catalog No. NC1130608 Supplier Fine Science Tools Supplier No. 9111710
Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.2 x 0.2mm/ Alloy / Material: Student Stainless Steel/ Length: 11cm/ Feature: Student Quality