Learn More
Fine Science Tools Student Fine Forceps, Straight, Stainless Steel, 11 cm
SDP
Catalog No. NC1045957
Encompass
Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.2 x 0.1mm/ Alloy / Material: Student Stainless Steel/ Length: 11cm/ Feature: Student Quality
Catalog No. NC1045957 Supplier Fine Science Tools Supplier No. 9111510
Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.2 x 0.1mm/ Alloy / Material: Student Stainless Steel/ Length: 11cm/ Feature: Student Quality