Learn More
Fine Science Tools Student Halsted-Mosquito Hemostats, Curved, 12.5 cm
SDP
Catalog No. NC9241884
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: / Tip Width: 1.4mm/ Clamping Length: 20mm/ Alloy / Material: Student Stainless Steel/ Length: 12.5cm/ Feature: Student Quality
Catalog No. NC9241884 Supplier Fine Science Tools Supplier No. 9130912
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: / Tip Width: 1.4mm/ Clamping Length: 20mm/ Alloy / Material: Student Stainless Steel/ Length: 12.5cm/ Feature: Student Quality