Learn More
World Precision Instrument Student Micro Surgery Scissors Curved Blades Flat Handle,15cm
SDP
Catalog No. 502538325
Encompass
Microsurgery scissors for students are budget-friendly compared to high-end surgical instruments. Student microsurgery scissors are made from stainless steel. They have straight or curved blades. The spring handles are designed for ease of use and perfect for right and left-handed users. They are flat and textured for a firm grasp. These scissors are suitable for educational environments where students are learning and practicing microsurgical techniques, or they may be used as single use instruments. They are autoclavable.
Catalog No. 50-253-8325 Supplier World Precision Instrument Supplier No. WP2800
Microsurgery scissors for students are budget-friendly compared to high-end surgical instruments. Student microsurgery scissors are made from stainless steel. They have straight or curved blades. The spring handles are designed for ease of use and perfect for right and left-handed users. They are flat and textured for a firm grasp. These scissors are suitable for educational environments where students are learning and practicing microsurgical techniques, or they may be used as single use instruments. They are autoclavable.