Learn More
Motic™ Swift Line Student Monocular LED Microscope M3604C-3
SCI_ED
Catalog No. S80701 Shop All Motic Swift Line Products
Change view
Click to view available options
Magnification Power:
40X, 100X, 400X
Catalog No. Magnification Power
S80701 40X, 100X, 400X
1 options
Catalog No. S80701 Supplier Motic™ Swift Line Supplier No. M3604C3
Add to Cart
Add to Cart
Built-in handle, variable LED illumination and lead-free objectives are just some of the state-of-the-art features that makes this series in a class of its own. Ideal for high school students and advanced grade levels.
OPTICAL SYSTEM
  • Widefield 10x /18 mm eyepiece.
  • Monocular viewing head with 45° inclined eyepiece.
  • DIN standard 4x (0.10 N.A.), 10x (0.25 N.A.), and 40xR (0.65 N.A.) achromat objectives. All are parfocalled, parcentered and color-coded.
  • Quad objective turret is ball-bearing mounted for smooth, precise positioning of objectives. Mounted in reverse position to facilitate ease in changing slides.
  • Camera port integrated into head for attachment of optional Moticam camera
  • C-mount lens and 1.5 mm wrench included
ILLUMINATION
  • LED light rated up to 50,000 hours of life, illumination equal to a 20 watt tungsten bulb.
  • Batteries allow up to 500 recharges, for a total of 20,000 to 25,000 hours use from a single set of 3 AA rechargeable nickel hydride batteries. Battery charger included with microscope, automatically shuts off when batteries are fully charged, no danger of overcharging batteries and shortening battery life. No expensive recharger station needed/ Plug directly into wall outlet, or use ordinary, inexpensive power strip for recharging multiple microscopes. Indicator light changes from red to green when fully charged.
  • Single 8 hour charge allows up to 50 hours of use.
ELECTRICAL
  • 110 V - 220 V switching power supply, unit shipped with American plug.
FOCUSING
  • Low position coarse and fine focusing controls.
  • Stage moves up or down, eyepiece and body remain fixed.
  • Slip clutch prevents damage to focusing system.
  • Tension adjustment eliminates stage drift.
  • Safety rack stop prevents damage to slides and objective lenses.
STAGE
  • Large 5-3/4 in. x 4-3/4 in. (143 mm x 120 mm) stage; built-in, low profile, ball-bearing mounted mechanical specimen holder with low-position coaxial controls.
  • Built-in 1.25 Abbe condenser has iris diaphragm.
Specifications
Head Style Monocular
Lighting Type LED
Eyepiece WF 10X/18 mm
Model M3604C-3
Product Type Monocular Microscope
Voltage 110/220 V
Magnification Power 40X, 100X, 400X
Objective Types Achromatic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov