Learn More
World Precision Instrument Student Noyes Iris Scissors Straight, 11.5 cm
SDP
Supplier:  World Precision Instrument WP2940
Encompass
Noyes Micro Scissors are a surgical instrument designed for ophthalmic surgery and are an ideal choice for microdissection or for delicate small animal surgeries. Named after their inventor, ophthalmologist Dr. Arthur Noyes, the Noyes Micro Scissors are characterized by their small size and fine tips. They let you make precise and controlled incisions. These surgical scissors are typically used for delicate tasks such as cutting fine structures, trimming tissue, or creating small openings. Noyes scissors are a type of spring scissors which can be used ambidextrously. For this reason, Noyes scissors are often preferred by left-handed surgeons. These scissors are 11.5 cm (4.5") long and come with 1.5 cm straight cutting edge and sharp/sharp tips. Noyes Micro Scissors are made from surgical grade stainless steel, which is suitable for autoclaving or chemical sterilization. These are a student grade instrument. Always clean your surgical instruments promptly after you use them.
Catalog No. 50-253-8323