Learn More
World Precision Instrument Student Operating Scissors with Tungsten Carbide ,14.5 cm
SDP
Catalog No. 502538343
Encompass
Ring-handled Operating Scissors are a pair of surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 14.5 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors have one sharp tip and one blunt one. They are available in surgical grade stainless steel with Tungsten Carbide inserts. They are durable, and the tungsten carbide inserts ensure they stay sharp longer. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. All surgical instruments should be thoroughly cleaned immediately after every use.
Catalog No. 50-253-8343 Supplier World Precision Instrument Supplier No. WP1112TC
Ring-handled Operating Scissors are a pair of surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 14.5 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors have one sharp tip and one blunt one. They are available in surgical grade stainless steel with Tungsten Carbide inserts. They are durable, and the tungsten carbide inserts ensure they stay sharp longer. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. All surgical instruments should be thoroughly cleaned immediately after every use.