Learn More
Surgical Design™ Student Otoscope Set With Replaceable Specula and Extra Bulb
Shop All Surgical Design ProductsDescription
- Full Student Otoscope Set including otoscope head, handle, extra bulb, and multi-size replaceable specula
- Includes 50-pack disposable specula for hygienic use across multiple examinations
- Replaceable specula sizes designed for pediatric and adult ear exams
- Color options: Standard, Black, or Blue
- Bright, clear halogen illumination ideal for learning environments and clinical use
- Durable construction suitable for medical students, nursing programs, EMT training, and at-home diagnostics
- Additional Specula Available for Purchase
Specifications
Specifications
| Voltage | 3 V |
| For Use With (Equipment) | Diagnostic Otoscopes |
| Product Type | Otoscope |
| Disposable | Reusable |
| Includes | Style: Blue Handle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.