Learn More
Fine Science Tools Student Surgical Scissors, Stainless Steel, Straight, 14.5 cm
SDP
Catalog No. NC9276852
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 45mm/ Alloy / Material: Student Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Student Quality
Catalog No. NC9276852 Supplier Fine Science Tools Supplier No. 9140214
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 45mm/ Alloy / Material: Student Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Student Quality