Learn More
Fine Science Tools Student Anatomical Standard Pattern Forceps, Stainless Steel, 16 cm
SDP
Catalog No. NC0445947
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 3.5 x 1.75mm/ Alloy / Material: Student Stainless Steel/ Length: 16cm/ Feature: Student Quality
Catalog No. NC0445947 Supplier Fine Science Tools Supplier No. 9110016
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 3.5 x 1.75mm/ Alloy / Material: Student Stainless Steel/ Length: 16cm/ Feature: Student Quality