Learn More
Fine Science Tools Student Surgical Scissors, Stainless Steel, Staight, 14.5 cm
SDP
Catalog No. NC1523660
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Student Stainless Steel/ Length: 14.5cm/ Serrated: No
Catalog No. NC1523660 Supplier Fine Science Tools Supplier No. 9140014
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Student Stainless Steel/ Length: 14.5cm/ Serrated: No