Learn More
Fine Science Tools Student Surgical Scissors, Stainless Steel, Staight, 12 cm
SDP
Catalog No. NC0144663
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 40mm/ Alloy / Material: Student Stainless Steel/ Length: 12cm/ Serrated: No/ Feature: Student Quality
Catalog No. NC0144663 Supplier Fine Science Tools Supplier No. 9140012
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 40mm/ Alloy / Material: Student Stainless Steel/ Length: 12cm/ Serrated: No/ Feature: Student Quality