Learn More
Fine Science Tools STUDENT VANNAS SPRING SCISSORS
SDP
Catalog No. NC9609583
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.35mm/ Alloy / Material: Student Stainless Steel/ Length: 9cm/ Serrated: No/ Feature: Student Quality
Catalog No. NC9609583 Supplier Fine Science Tools Supplier No. 9150009
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.35mm/ Alloy / Material: Student Stainless Steel/ Length: 9cm/ Serrated: No/ Feature: Student Quality