Learn More
Teknova Studier ZYM-5052 Medium Kit. 1L Kit
SDP
Catalog No. NC1093977
Encompass
Studier ZYM-5052 Medium Kit, To complete media aseptically add 60mL of 3S2600 to 3S2200, Media is stable 2 months after salts are added
Catalog No. NC1093977 Supplier Teknova Supplier No. 3S2000
Studier ZYM-5052 Medium Kit, To complete media aseptically add 60mL of 3S2600 to 3S2200, Media is stable 2 months after salts are added