Learn More
Teknova Studier ZYP-5052 Medium Kit. 1L Kit
SDP
Catalog No. NC3315082
Encompass
Studier ZYP-5052 Medium Kit, To complete media aseptically add 120mL of 3S8600 to 3S8200
Catalog No. NC3315082 Supplier Teknova Supplier No. 3S8000
Studier ZYP-5052 Medium Kit, To complete media aseptically add 120mL of 3S8600 to 3S8200