Learn More
New England Biolabs, Inc. StuI – 1000 units
SDP
Catalog No. 50812106
Encompass
A restriction endonuclease that recognizes the sequence AGG_^CCT.
Catalog No. 50-812-106 Supplier New England Biolabs, Inc. Supplier No. R0187S
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence AGG_^CCT.