Learn More
New England Biolabs, Inc. StuI – 5000 units
SDP
Catalog No. 50812104
Encompass
A restriction endonuclease that recognizes the sequence AGG_^CCT.
Catalog No. 50-812-104 Supplier New England Biolabs, Inc. Supplier No. R0187L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence AGG_^CCT.