Learn More
Unchained Labs Stunner, Unchained Labs
SDP
Catalog No. NC1767970
Encompass
The Unchained Labs Stunner is a high-performance benchtop biophysical characterization system for proteins. It uses multi-wavelength detection to measure size, structure, stability and interactions. Key features include native mass detection up to 1 MDa, reversible photochemical unfolding, and label-free binding kinetics. The Stunner enables rapid screening and ranking of protein therapeutics earlier in development to derisk biologics pipelines. Its applications include candidate screening, comparability studies, formulation studies and protein engineering. The system provides automation, throughput and ease-of-use.
This item is not returnable. View return policy
Catalog No. NC1767970 Supplier Unchained Labs Supplier No. 7012022
The Unchained Labs Stunner is a high-performance benchtop biophysical characterization system for proteins. It uses multi-wavelength detection to measure size, structure, stability and interactions. Key features include native mass detection up to 1 MDa, reversible photochemical unfolding, and label-free binding kinetics. The Stunner enables rapid screening and ranking of protein therapeutics earlier in development to derisk biologics pipelines. Its applications include candidate screening, comparability studies, formulation studies and protein engineering. The system provides automation, throughput and ease-of-use.