Learn More
C.L. Sturkey Scissors Sharpening Service
Catalog No. 22210031 Shop All CL Sturkey Products
Catalog No. 22-210031 Supplier C.L. Sturkey Supplier No. S100
Add to Cart
Add to Cart
Sharpening service for scissors.
Service for:
  • Premium blades: durable and economical with uncoated cutting edges; for general paraffin and cryostat use.
  • Silver blades: PTFE resin coated cutting edge; for use in general paraffin sectioning when ultra-thin sections are required.
  • Gold blades: edges are coated with titanium nitride that adds durability, versatility and longevity to the cutting edge; ideal for general paraffin and cryostat use.
  • Diamond blades: diamond-coated and excellent for general paraffin sectioning with special effectiveness on hard tissues and decalcified bones; cutting edge is more durable and lasts two to three times longer than conventional blades.
Specifications
Product Type Sharpening Service