Learn More
World Precision Instrument STX HTS EVOM Electrode for CORNING 24
SDP
Catalog No. 502521183
Encompass
Semi-permeable HTS (high throughput screening) culture plates have become a standard tool for pharmaceutical and institutional research in epithelial transport. HTS culture plates have well cups which are bonded together into a single assembly and are not removable. This makes the plates ideal for automated applications, but imposes a significant inconvenience when TEER must be measured in the absence of an automated system. WPI’s STX HTS series of EVOM™ electrodes, replacing the STX100 electrodes, provide a cost-effective alternative to automation, allowing HTS well plates to be measured manually using a hand–held electrode.
To learn more about our warranty options, click here.
Catalog No. 50-252-1183 Supplier World Precision Instrument Supplier No. EVMEL030304
Semi-permeable HTS (high throughput screening) culture plates have become a standard tool for pharmaceutical and institutional research in epithelial transport. HTS culture plates have well cups which are bonded together into a single assembly and are not removable. This makes the plates ideal for automated applications, but imposes a significant inconvenience when TEER must be measured in the absence of an automated system. WPI’s STX HTS series of EVOM™ electrodes, replacing the STX100 electrodes, provide a cost-effective alternative to automation, allowing HTS well plates to be measured manually using a hand–held electrode.
To learn more about our warranty options, click here.