Learn More
Aviva Systems Biology Corp STX19 Antibody - N-terminal region (ARP56145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STX19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STX19. Alternate names: MGC21382ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-61737