Learn More
Aviva Systems Biology Corp Stx1a Antibody - N-terminal region (ARP59486_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59486_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Stx1a. It was validated on Western Blot. . Alternate names: HPC-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-715