Learn More
Aviva Systems Biology Corp STX1A Antibody - N-terminal region (AVARP13091_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13091_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STX1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STX1A. Alternate names: HPC-1, STX1, p35-1, P35-1, SYN1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-62638