Learn More
Aviva Systems Biology Corp STX2 Antibody - N-terminal region (ARP61202_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61202_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STX2. It was validated on Western Blot. . Alternate names: EPIM, EPM, MGC51014, STX2A, STX2B, STX2CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-292