Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STX7 Human HEK293 10ug
SDP

Catalog No. 5000196846
Encompass_Preferred

Recombinant human STX7 (syntaxin-7) supplied as a purified His-tagged protein produced in HEK293 cells, designed for biochemical and functional studies where native-like folding and post-translational modifications are important. The preparation has low endotoxin and is provided with storage and handling recommendations for long-term stability.

  • Human species, sequence range S2-L238 (Uniprot O15400).
  • Expressed in HEK293 mammalian cells for authentic folding and modifications.
  • Contains a His tag to facilitate purification and detection.
  • Molecular weight approximately 31-36 kDa.
  • Endotoxin level <1 EU/μg, determined by LAL method.
  • Store at -20°C; stable at 4°C for one week after reconstitution; aliquot and freeze at -20°C or -80°C for long-term storage.
  • Suitable for antibody validation, biochemical assays, and functional studies.

Catalog No. 50-001-96846 Supplier Medchemexpress LLC Supplier No. HYP7102410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human STX7 (syntaxin-7) supplied as a purified His-tagged protein produced in HEK293 cells, designed for biochemical and functional studies where native-like folding and post-translational modifications are important. The preparation has low endotoxin and is provided with storage and handling recommendations for long-term stability.

  • Human species, sequence range S2-L238 (Uniprot O15400).
  • Expressed in HEK293 mammalian cells for authentic folding and modifications.
  • Contains a His tag to facilitate purification and detection.
  • Molecular weight approximately 31-36 kDa.
  • Endotoxin level <1 EU/μg, determined by LAL method.
  • Store at -20°C; stable at 4°C for one week after reconstitution; aliquot and freeze at -20°C or -80°C for long-term storage.
  • Suitable for antibody validation, biochemical assays, and functional studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.