Learn More
Aviva Systems Biology Corp STXBP1 Antibody - middle region (AVARP13108_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13108_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STXBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human STXBP1. Alternate names: MUNC18-1, UNC18, hUNC18, rbSec1, P67, NSEC1, RBSEC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Zebrafish
Catalog No. 50-014-62643