Learn More
Aviva Systems Biology Corp STXBP4 Antibody - N-terminal region (ARP65839_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65839_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STXBP4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human STXBP4. Alternate names: FLJ16496, MGC149829, MGC50337, SynipApplicationWB. Species reactivity: Human
Catalog No. 50-288-578