Learn More
Aviva Systems Biology Corp Stxbp6 Antibody - N-terminal region (ARP54989_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Stxbp6. It was validated on Western Blot. . Alternate names: BC024598, C85317, MGC30948ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-986