Learn More
New England Biolabs, Inc. StyD4I – 200 units
SDP
Catalog No. 50812390
Encompass
A restriction endonuclease that recognizes the sequence ^CCNGG_.
Catalog No. 50-812-390 Supplier New England Biolabs, Inc. Supplier No. R0638S
A restriction endonuclease that recognizes the sequence ^CCNGG_.