Learn More
Aviva Systems Biology Corp STYK1 Antibody - C-terminal region (ARP47449_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47449_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STYK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human STYK1. Alternate names: DKFZp761P1010, NOK, SuRTK106ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59554