Learn More
Electron Microscopy Sciences EMS Style 4WF PVDF Replacement Tips, OAL
SDP
Catalog No. 501930874
Encompass_Preferred
Durable self-aligning replacement PVDF tips and screws for replaceable tip tweezers Style 4WF. Tip with stepped bottom paddle and top fingers for handling 2 inch wafers.
Material: PVDF
Catalog No. 50-193-0874 Supplier Electron Microscopy Sciences Supplier No. 7832562
Durable self-aligning replacement PVDF tips and screws for replaceable tip tweezers Style 4WF. Tip with stepped bottom paddle and top fingers for handling 2 inch wafers.
Material: PVDF