Learn More
Electron Microscopy Sciences Rubis Style Micro Scissors, 1C300
SDP
Catalog No. 50364881
Encompass_Preferred
High quality scissors, manufactured from hardened, anti-acid, stainless, surgical steel (CA) Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist grip scissors offer ergonomically, comfort and control. Sharp pointed micro scissors.
OAL: 110 mm
Size: N = 5.4 mm
Catalog No. 50-364-881 Supplier Electron Microscopy Sciences Supplier No. 781801C3
High quality scissors, manufactured from hardened, anti-acid, stainless, surgical steel (CA) Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist grip scissors offer ergonomically, comfort and control. Sharp pointed micro scissors.
OAL: 110 mm
Size: N = 5.4 mm