Learn More
Med Vet International Dynarex intubating Stylettes, 12 Fr, 20/Box
Catalog No. 502831442
Encompass_Preferred
Dynarex Resp-O2™ Intubating Stylettes are designed to be used in conjunction with endotracheal tubes to provide a smoother, more efficient intubation process. These premium-quality Stylettes are engineered with a malleable aluminum wire to support and guide the endotracheal tube into the patient’s airway. The Stylette is encased in a flexible plastic sheath that extends beyond the aluminum wire to help reduce friction and minimize the risk of tracheal trauma. Dynarex Resp-O2™ Intubating Stylettes are available in a variety of sizes to fit most adult and pediatric tracheal tubes.  Dynarex Resp-O2™ Intubating Stylettes are designed to facilitate safe and effective endotracheal intubations. The Stylettes feature a malleable aluminum wire encased in a plastic sheath to help reduce friction for smoother insertion and removal of endotracheal tubes. 
Catalog No. 50-283-1442 Supplier Med Vet International Supplier No. 36267
Dynarex Resp-O2™ Intubating Stylettes are designed to be used in conjunction with endotracheal tubes to provide a smoother, more efficient intubation process. These premium-quality Stylettes are engineered with a malleable aluminum wire to support and guide the endotracheal tube into the patient’s airway. The Stylette is encased in a flexible plastic sheath that extends beyond the aluminum wire to help reduce friction and minimize the risk of tracheal trauma. Dynarex Resp-O2™ Intubating Stylettes are available in a variety of sizes to fit most adult and pediatric tracheal tubes.  Dynarex Resp-O2™ Intubating Stylettes are designed to facilitate safe and effective endotracheal intubations. The Stylettes feature a malleable aluminum wire encased in a plastic sheath to help reduce friction for smoother insertion and removal of endotracheal tubes. 