Learn More
Waters Corp Waters Styragel GPC Guard Column, 20 μm, 4.6 mm, 30 mm, DMF
Catalog No. 50785716 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel Guard columns increase the lifetime of 7.8 mm or 4.6 mm ID columns. This GPC Guard Column protects GPC Columns in reversed-phase Liquid Chromatography by filtering chemical and particulate impurities from the mobile phase stream, enhancing analytical performance.
  • Manufactured under cGMP, ISO 9001 conditions, ensuring optimal performance, selectivity, sensitivity, and speed of analysis.
  • Testing results meet high standards and narrow specification ranges for excellent, reproducible performance.
  • Produced using premier materials, enhancing the GPC columns\' performance and longevity.
  • Removes contaminants, maintaining UPLC, UHPLC, and HPLC separation efficiency.
Catalog No. 50-785-716 Supplier Waters Corp Supplier No. WAT054415
Styragel Guard columns increase the lifetime of 7.8 mm or 4.6 mm ID columns. This GPC Guard Column protects GPC Columns in reversed-phase Liquid Chromatography by filtering chemical and particulate impurities from the mobile phase stream, enhancing analytical performance.
  • Manufactured under cGMP, ISO 9001 conditions, ensuring optimal performance, selectivity, sensitivity, and speed of analysis.
  • Testing results meet high standards and narrow specification ranges for excellent, reproducible performance.
  • Produced using premier materials, enhancing the GPC columns\' performance and longevity.
  • Removes contaminants, maintaining UPLC, UHPLC, and HPLC separation efficiency.