Learn More
Waters Corp Waters Styragel Guard Column, 20 μm, 4.6 mm X 30 mm, THF
Catalog No. 50785714 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel Guard Columns are designed to protect and extend the lifetime of analytical columns, especially those with 7.8 mm or 4.6 mm ID. They are suitable for high-resolution analysis of oligomers, epoxies, polymer additives, and low molecular weight samples. These guard columns are manufactured under strict ISO, FDA, and cGMP guidelines.
  • Protects analytical columns and extends their lifetime.
  • Ideal for high-resolution analysis of oligomers, epoxies, and polymer additives.
  • Designed for use with low molecular weight samples.
  • Manufactured under strict ISO, FDA, and cGMP guidelines for reproducible technology.
  • Helps prevent buildup in system operating pressure caused by contamination.
Catalog No. 50-785-714 Supplier Waters Corp Supplier No. WAT054405
Styragel Guard Columns are designed to protect and extend the lifetime of analytical columns, especially those with 7.8 mm or 4.6 mm ID. They are suitable for high-resolution analysis of oligomers, epoxies, polymer additives, and low molecular weight samples. These guard columns are manufactured under strict ISO, FDA, and cGMP guidelines.
  • Protects analytical columns and extends their lifetime.
  • Ideal for high-resolution analysis of oligomers, epoxies, and polymer additives.
  • Designed for use with low molecular weight samples.
  • Manufactured under strict ISO, FDA, and cGMP guidelines for reproducible technology.
  • Helps prevent buildup in system operating pressure caused by contamination.