Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Styragel Column, HMW 2, 500 Å, 20 μm

Catalog No. 50785740 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HMW columns are designed for high-molecular weight applications, featuring a 20 μm particle and a specialized 10 μm frit to minimize polymer shearing.

  • Packed with a 20 μm particle
  • Utilizes a special 10 μm frit to minimize polymer shearing
  • Engineered for high-molecular weight applications
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 100 to 10000
  • Spherical particle shape
  • Pore size: 500 Å
  • Column format
  • Packing solvent: Toluene
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • 1 unit per package

Catalog No. 50-785-740 Supplier Waters Corp Supplier No. WAT054490
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HMW columns are designed for high-molecular weight applications, featuring a 20 μm particle and a specialized 10 μm frit to minimize polymer shearing.

  • Packed with a 20 μm particle
  • Utilizes a special 10 μm frit to minimize polymer shearing
  • Engineered for high-molecular weight applications
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 100 to 10000
  • Spherical particle shape
  • Pore size: 500 Å
  • Column format
  • Packing solvent: Toluene
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • 1 unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.