Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel Column, HMW 2, 500 Å, 20 μm, 7.8 mm × 300 mm

Catalog No. 50785739 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HMW columns are packed with a 20 μm particle and utilize a special 10 μm frit to minimize polymer shearing, making them ideal for use in high-molecular weight applications.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 100 to 10000
  • Particle shape: Spherical
  • Particle size: 20 μm
  • Endfitting type: Standard
  • Pore size: 500 Å
  • Format: Column
  • Packing solvent: THF
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1 pk

Catalog No. 50-785-739 Supplier Waters Corp Supplier No. WAT054488
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HMW columns are packed with a 20 μm particle and utilize a special 10 μm frit to minimize polymer shearing, making them ideal for use in high-molecular weight applications.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 100 to 10000
  • Particle shape: Spherical
  • Particle size: 20 μm
  • Endfitting type: Standard
  • Pore size: 500 Å
  • Format: Column
  • Packing solvent: THF
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.