Learn More
Waters Corp WATERS CORPORATION Styragel HMW 6E Column, 5,000 to 1 x 10^7 Molecular Weight, Toluene
Catalog No. 50785525 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Styragel HMW 6E Column is designed for the analysis of ultra-high-molecular-weight polymers. This long-life column is packed with 20 μm particles and is suitable for use at both ambient and elevated temperatures. It features an effective molecular weight range of 5,000 to 1 x 107 and is shipped in Toluene.
  • Designed for the analysis of ultra-high-molecular-weight polymers.
  • Packed with 20 μm particles.
  • Suitable for use at both ambient and elevated temperatures.
  • Features an effective molecular weight range of 5,000 to 1 x 107.
  • Shipped in Toluene.
Catalog No. 50-785-525 Supplier Waters Corp Supplier No. WAT044203
The Styragel HMW 6E Column is designed for the analysis of ultra-high-molecular-weight polymers. This long-life column is packed with 20 μm particles and is suitable for use at both ambient and elevated temperatures. It features an effective molecular weight range of 5,000 to 1 x 107 and is shipped in Toluene.
  • Designed for the analysis of ultra-high-molecular-weight polymers.
  • Packed with 20 μm particles.
  • Suitable for use at both ambient and elevated temperatures.
  • Features an effective molecular weight range of 5,000 to 1 x 107.
  • Shipped in Toluene.