Learn More
Waters Corp Waters Styragel HMW 6E Column, 20 μm, 7.8 mm x 300 mm
Supplier:  Waters Corp WAT044205
Encompass_Preferred
Styragel HMW columns are packed with a 20 μm particle and utilize a special 10 μm frit. This design is intended to minimize polymer shearing, making them suitable for use in high-molecular weight applications.
  • Utilizes a special 10 μm frit to minimize polymer shearing.
  • Ideal for use in high-molecular weight applications.
Catalog No. 50-785-527