Learn More
Waters Corp Waters Column, 15–20 μm, 7.8 x 300 mm, Mixed Bed
Supplier: Waters Corp WAT044204
Styragel HMW columns are designed for high-molecular weight applications, utilizing a 20 μm particle and 10 μm frit to minimize polymer shearing. They ensure high quality and desired results for liquid chromatography, offering exceptional longevity at various temperatures.
- Separation mode: SEC/GPC
- Polymer particle substrate
- pH range: 2 to 12
- Molecular weight range: 5,000 to 10,000,000
- Spherical particle shape
- Particle size: 15 to 20 μm
- Mixed pore size
- Packing solvent: THF
- System: HPLC
- Technique: GPC
- Inner diameter: 7.8 mm
- Length: 300 mm
- Units per package: 1
- Minimizes polymer shearing
- Exceptional column longevity
- Usable at ambient or increased temperatures
- Manufactured to ISO, FDA, and cGMP standards
- Ensures high column efficiency and repeatability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.