Learn More
Waters Corp Waters Styragel Column, HMW 6E, Mixed Bed, 20 μm, 4.6 mm X 300 mm, Toluene
Supplier:  Waters Corp WAT046815
Encompass_Preferred
These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. Styragel HMW columns are packed with a 20 μm particle and utilize a special 10 μm frit to minimize polymer shearing, making them ideal for use in high-molecular weight applications.
  • Reduced solvent consumption and disposal costs by up to two-thirds.
  • Minimized polymer shearing with a 20 μm particle and special 10 μm frit.
  • Ideal for high-molecular weight applications.
Catalog No. 50-785-620