Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel HMW 7 Column, 15 to 20 μm, 4.6 mm X 300 mm

Catalog No. 50785617 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. They are packed with a 20 μm particle and utilize a special 10 μm frit to minimize polymer shearing, making them ideal for use in high-molecular weight applications.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 500,000 to 100,000,000
  • Particle shape: Spherical
  • Particle size: 15 to 20 μm
  • Pore size: 10,000,000 Å
  • Packing solvent: Toluene
  • System: HPLC
  • Inner diameter: 4.6 mm
  • Length: 300 mm
  • Designed to reduce solvent consumption and disposal costs
  • Incorporates a special 10 μm frit to minimize polymer shearing
  • Suitable for high-molecular weight applications

Catalog No. 50-785-617 Supplier Waters Corp Supplier No. WAT046800
May include imposed supplier surcharges.
Add to Cart
Add to Cart

These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. They are packed with a 20 μm particle and utilize a special 10 μm frit to minimize polymer shearing, making them ideal for use in high-molecular weight applications.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 500,000 to 100,000,000
  • Particle shape: Spherical
  • Particle size: 15 to 20 μm
  • Pore size: 10,000,000 Å
  • Packing solvent: Toluene
  • System: HPLC
  • Inner diameter: 4.6 mm
  • Length: 300 mm
  • Designed to reduce solvent consumption and disposal costs
  • Incorporates a special 10 μm frit to minimize polymer shearing
  • Suitable for high-molecular weight applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.