Learn More
Waters Corp Waters Styragel HMW 7 Column, 20 μm, 4.6 mm ID x 300 mm L, DMF
Catalog No. 50785619 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HMW 7 columns are designed for the analysis of ultra-high-molecular-weight polymers, which are often susceptible to shearing during analysis. This specific column features a 20 μm particle size, 4.6 mm inner diameter, and 300 mm length, and is shipped in Dimethylformamide (DMF) for immediate use in relevant applications.
  • Designed to analyze polymers of ultra-high-molecular-weight that are susceptible to shearing
  • Long-life column
  • Suitable for ultra-high molecular weight analysis
  • Applications include UHDPE, UH-Polystyrene, UH-Isoprene, and UH-PMMA
  • Suitable for use at both ambient and elevated temperatures
  • Effective molecular weight range of 500,000 to 1 x 108
Catalog No. 50-785-619 Supplier Waters Corp Supplier No. WAT046810
Styragel HMW 7 columns are designed for the analysis of ultra-high-molecular-weight polymers, which are often susceptible to shearing during analysis. This specific column features a 20 μm particle size, 4.6 mm inner diameter, and 300 mm length, and is shipped in Dimethylformamide (DMF) for immediate use in relevant applications.
  • Designed to analyze polymers of ultra-high-molecular-weight that are susceptible to shearing
  • Long-life column
  • Suitable for ultra-high molecular weight analysis
  • Applications include UHDPE, UH-Polystyrene, UH-Isoprene, and UH-PMMA
  • Suitable for use at both ambient and elevated temperatures
  • Effective molecular weight range of 500,000 to 1 x 108