Learn More
Waters Corp Waters Column, 15 - 20 μm, 7.8 mm × 300 mm, 10,000,000 Å Pore
Catalog No. 50785523 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HMW columns are engineered for high-molecular-weight applications, featuring a 20 μm particle and a specialized 10 μm frit to minimize polymer shearing. They are developed for analyzing ultra-high molecular weight polymers, offering significant column durability and reliable results for liquid chromatography.
  • Minimizes polymer shearing due to specialized frit and particle size.
  • Ideal for high-molecular-weight applications.
  • Suitable for analyzing ultra-high molecular weight polymers.
  • Can be employed at ambient or elevated temperatures.
  • Offers remarkable column durability.
  • Manufactured adhering to ISO, FDA, and cGMP standards.
  • Delivers high quality and desired results for liquid chromatography.
  • Provides crucial performance traits: column efficiency, peak symmetry, column longevity, and column-to-column repeatability.
Catalog No. 50-785-523 Supplier Waters Corp Supplier No. WAT044201
Styragel HMW columns are engineered for high-molecular-weight applications, featuring a 20 μm particle and a specialized 10 μm frit to minimize polymer shearing. They are developed for analyzing ultra-high molecular weight polymers, offering significant column durability and reliable results for liquid chromatography.
  • Minimizes polymer shearing due to specialized frit and particle size.
  • Ideal for high-molecular-weight applications.
  • Suitable for analyzing ultra-high molecular weight polymers.
  • Can be employed at ambient or elevated temperatures.
  • Offers remarkable column durability.
  • Manufactured adhering to ISO, FDA, and cGMP standards.
  • Delivers high quality and desired results for liquid chromatography.
  • Provides crucial performance traits: column efficiency, peak symmetry, column longevity, and column-to-column repeatability.