Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters GPC Column, Styragel HR 0.5, 5 μm, 4.6 mm ID, 300 mm

Catalog No. 50785586 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These narrow bore columns reduce solvent consumption and disposal costs. They are high-resolution GPC columns with 5 μm particles, ideal for characterizing samples in the low- to mid-molecular range.

  • Reduces solvent consumption and disposal costs.
  • High-resolution GPC packed with 5 μm particles.
  • Characterizes samples in low- to mid-molecular weight range.
  • Separates molecules from interfering matrices in food, pharma, and natural goods.
  • Manufactured in cGMP, ISO 9001-certified facility.
  • Chromatographically tested for repeatable performance.
  • Performance chromatogram and certificate of batch analysis on eCord™ Intelligent Chip.
  • Broad operating pH range of 2 to 12.
  • Effective molecular weight range of 0 to 1,000.
  • Dimensions: 4.6 mm ID x 300 mm length.
  • Pore size: 50 Å.
  • Packing solvent: THF.

Catalog No. 50-785-586 Supplier Waters Corp Supplier No. WAT045835
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These narrow bore columns reduce solvent consumption and disposal costs. They are high-resolution GPC columns with 5 μm particles, ideal for characterizing samples in the low- to mid-molecular range.

  • Reduces solvent consumption and disposal costs.
  • High-resolution GPC packed with 5 μm particles.
  • Characterizes samples in low- to mid-molecular weight range.
  • Separates molecules from interfering matrices in food, pharma, and natural goods.
  • Manufactured in cGMP, ISO 9001-certified facility.
  • Chromatographically tested for repeatable performance.
  • Performance chromatogram and certificate of batch analysis on eCord™ Intelligent Chip.
  • Broad operating pH range of 2 to 12.
  • Effective molecular weight range of 0 to 1,000.
  • Dimensions: 4.6 mm ID x 300 mm length.
  • Pore size: 50 Å.
  • Packing solvent: THF.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.